Learn More
Novus Biologicals™ Fixation and Permeabilization Flow Cytometry Kit
Product Code. 30519911 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
200 mL (2 x 100 mL)
8 x 5 mL
Unit Size:
1 set
Product Code. Quantity unitSize
30519911 200 mL (2 x 100 mL) 1 set
30519912 8 x 5 mL 1 set
2 options
Product Code. 30519911
Brand: Novus Biologicals™ NBP231377200ml
Intended for first fixing cells in suspension with Reagent A and then permeabilizing the cell membranes with Reagent B
TRUSTED_SUSTAINABILITY
Specifications
Content And Storage Store at room temperature.
For Use With (Application) Flow Cytometry
Quantity 200 mL (2 x 100 mL)