Learn More
FITM, MedChemExpress
Product Code. 16484696
Encompass_Preferred
Change view
Click to view available options
Quantity:
2 mg
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
2mg
50mg
5mg
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16484696 100 mg 100mg
16463608 10 mg 10mg
16462786 25 mg 25mg
16436337 5 mg 5mg
16430066 2 mg 2mg
16400007 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Product Code. 16484696 Supplier Medchem Express Supplier No. HY101845
MedChemExpress FITM is a negative allosteric modulator of mGlu1 receptor with a Ki of 2.5 nM.
FITM is a negative allosteric modulator of mGlu1 receptor with a Ki of 2.5 nM.
Specifications
Chemical Name or Material FITM
CAS 932737-65-0
Color Off-White
Purity Grade Notes Research
Molecular Formula C18H18FN5OS
Quantity 100 mg
Solubility Information DMSO : ≥ 150 mg/mL (403.84 mM)
SMILES O=C(N(C)C1=NC(C2=NC=NC(NC(C)C)=C2)=CS1)C3=CC=C(F)C=C3
Molecular Weight (g/mol) 371.43
Formula Weight 371.43
Percent Purity 98.19%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death