Learn More
Fisherbrand™ Universal Pipet Pump
Product Code. 15239805 Shop All Fisherbrand Traceable Products
Change view
Click to view available options
Color:
Blue
Green
Red
Volume (Metric):
2 mL
10 mL
25 mL
Unit Size:
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Volume (Metric) Unit Size
15239805 Green 10 mL Each
15229805 Red 25 mL Each
15209805 Blue 2 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 15239805
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Pipet filler with easy one-hand operation provides accurate titration. Color-coded for easy identification.
  • Made from high-grade polyethylene
  • Accepts glass and serological pipets
  • Ideal for soap and acid titrations
  • Clear markings make measuring easy
  • 10mL pipet marked in 0.10mL increments
  • 25mL pipet marked in 0.20mL increments
  • Rotating thumbwheel draws or dispensed liquid
  • Side lever allows entire contents to be rapidly dispensed
25mL fits both 10mL and 25mL pipets
Specifications
Color Green
Material Polyethylene
Type Pipet Controller
Volume (Metric) 10 mL