Learn More
Fisherbrand™ Universal Pipet Pump
Product Code. 15209805 Shop All Fisherbrand Traceable Products
Change view
Click to view available options
Color:
Blue
Green
Red
Volume (Metric):
2 mL
10 mL
25 mL
Unit Size:
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Volume (Metric) Unit Size
15209805 Blue 2 mL Each
15239805 Green 10 mL Each
15229805 Red 25 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
You may only order up to the available stock. For more details call customer services on +358 9 8027 6280.
Product Code. 15209805
Estimated Shipment: 12-01-2027
Add to basket
Add to basket
Bulk Quote Request
You may only order up to the available stock. For more details call customer services on +358 9 8027 6280.
Pipet filler with easy one-hand operation provides accurate titration. Color-coded for easy identification.
  • Made from high-grade polyethylene
  • Accepts glass and serological pipets
  • Ideal for soap and acid titrations
  • Clear markings make measuring easy
  • 10mL pipet marked in 0.10mL increments
  • 25mL pipet marked in 0.20mL increments
  • Rotating thumbwheel draws or dispensed liquid
  • Side lever allows entire contents to be rapidly dispensed
25mL fits both 10mL and 25mL pipets
Specifications
Color Blue
Material Polyethylene
Type Pipet Controller
Volume (Metric) 2 mL