Learn More
Fisherbrand™ Unbreakable Cover Slips
Product Code. 10452765 Shop All Fisherbrand Products
Change view
Click to view available options
Cover slip; Size: 22 x 22mm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
10452765 Cover slip; Size: 22 x 22mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10452765
Estimated Shipment: 28-08-2026
Add to basket
Add to basket
Plastic; clear, noncurling
  • Plastic (pressed poly vinyl chloride)
  • Clear
  • Noncurling
  • 0.17 to 0.25mm thick
Order Info
Pack of 1000 cover slips in boxes of 100
Specifications
Product Type Coverslip
Material Pressed PVC
Length (Metric) 22 mm
Width (Metric) 22 mm
Thickness (Metric) 0.17 to 0.25 mm
Quantity Ea.