Learn More
Fisherbrand™ Soil Analysis ASTM Hydrometers
Product Code. 17112435 Shop All Fisherbrand Traceable Products
Change view
Click to view available options
ASTM Number:
151H
Specific Gravity Range:
0.995 to 1.038
Accuracy:
±0.001
Increments:
0.001
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. ASTM Number Specific Gravity Range Accuracy Increments Unit Size
17112435 151H 0.995 to 1.038 ±0.001 0.001 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 17112435
  • ONLINE EXCLUSIVE PRICE
    81.90 EUR
    67.00 EUR / Pack of 1
    Save 14.90 EUR
    18% Off
Estimated Shipment: 18-12-2026
Add to basket
Add to basket
These hydrometers are used to determine the specific gravity of soil in the range of 0.995 to 1.038 or -5 to 60g/L (using a weight by volume scale).
  • 280mm (11 in.) long
  • Lead-free ballast and glass
  • Individually serialized
  • Conform to the accuracy requirements of ASTM hydrometers
  • Temperature standardization: 68°F
Storage:
Soil analysis hydrometers should be stored in a clean, dry, safe place, and should not be subjected to extreme temperatures.
Order Info
Should you require an individual Hydrometer Calibration Report, factory calibrated hydrometers or custom calibration services are available.
Certifications
Packaged with a multi-language (English, French, German, Italian, Portuguese, and Spanish) Statement of Accuracy indicating accuracy traceable to NIST. Hand Inspected by our U.S. quality team.
Specifications
ASTM Number 151H
Accuracy ±0.001
Increments 0.001
Certifications/Compliance Traceable to NIST
Length (Metric) 280 mm
Content And Storage Soil analysis hydrometers should be stored in a clean, dry, safe place, and should not be subjected to extreme temperatures.
For Use With (Application) To determine the specific gravity of soil in the range of 0.995 to 1.038 or -5 to 60g/L (using a weight by volume scale)
Specific Gravity Range 0.995 to 1.038