Learn More
Fisherbrand™ Screw Caps for Disposable Glass Tubes
Product Code. 12337279 Shop All Fisherbrand Traceable Products
Change view
Click to view available options
Color:
Black
Closure Size:
13-415
15-415
18-415
Material:
Phenolic
Unit Size:
Pack of 1000
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Closure Size Material Unit Size
12337279 Black 15-415 Phenolic Pack of 1000
11547133 Black 13-415 Phenolic Pack of 1000
11567133 Black 18-415 Phenolic Pack of 1000
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 12337279
Estimated Shipment: 29-12-2026
Add to basket
Add to basket
Phenolic caps have cemented-in rubber liners
  • Autoclavable
  • Polypropylene cap is linerless
Specifications
Closure Size 15-415
Liner Material Rubber
Pre-sterilized No
Product Type Screw Cap
Autoclavable Autoclavable
Material Phenolic
Color Black
Tip Material Rubber
Sterility Non-sterile
For Use With (Equipment) 14-959-35A, 14-959-35B, 14-959-35AA