Learn More
Fisherbrand™ Mini-Centrifuge Replacement Parts
Product Code. 12893332 Shop All Fisherbrand Traceable Products
Change view
Click to view available options
Capacity:
2 x 8 Strip 0.2 mL Tubes
6 x 1.5/2 mL
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity Unit Size
12893332 2 x 8 Strip 0.2 mL Tubes Each
12883332 6 x 1.5/2 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 12893332
  • ONLINE EXCLUSIVE PRICE
    57.85 EUR
    43.65 EUR / Pack of 1
    Save 14.20 EUR
    24% Off
 Restricted Item
Estimated Shipment: 17-12-2026
Add to basket
Add to basket
Replacement parts for Fisherbrand™ mini-centrifuge.
  • For use with the Fisherbrand™ mini-centrifuge
Specifications
Description Mini-centrifuge Rotor SnapSpin 2 x 8 Strip 0.2 mL
Material Polycarbonate
Type Rotor
Capacity 2 x 8 Strip 0.2 mL Tubes
Max. RCF None
Max. Speed None
For Use With (Equipment) Fisherbrand Mini-centrifuge
Biocontainment No