Learn More
Humidity Sponge™, Fisherbrand™
Shop All Cole Parmer Products
Click to view available options
Chemical Name or Material:
Fisherbrand™ Humidity Sponge™ Dessicant
Unit Size:
Pack of 40
Description
Humidity Sponge Dessicant; Fisherbrand; Reusable Type
- Non-toxic desiccant
- Reusable
- Material is made of DuPont's Tyvek™ breathable, high-density polyethylene
- Perfect for drugs, food, electronics, optics and equipment
- Bags are guaranteed to retain 16% moisture by weight, typical results are 37% moisture absorption
- Desiccant meets US FDA requirements
- Certified ISO Class 5 (Class M3.5) cleanroom compatible
Specifications
Specifications
| Chemical Name or Material | Fisherbrand™ Humidity Sponge™ Dessicant |
| CAS | 1302-78-9 |
| Material | HDPE |
| Quantity | 40 Pk. |
| Grade | Certified |
| Weight | 11 g |