Learn More
Fisherbrand™ Electroporation Cuvettes Plus™
Product Code. 15542423 Shop All Fisherbrand Traceable Products
Change view
Click to view available options
Capacity (Metric):
90 μL
400 μL
800 μL
Width (Metric) Gap:
1 mm
2 mm
4 mm
Unit Size:
Pack of 50
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Width (Metric) Gap Unit Size
15542423 400 μL 2 mm Pack of 50
15532423 90 μL 1 mm Pack of 50
15552423 800 μL 4 mm Pack of 50
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 15542423
Save Up to 60% on Microbiology Essentials
Use promo code 28004
View all promotions
Estimated Shipment: 01-12-2026
Add to basket
Add to basket
Disposable, color-coded cuvets fit most electroporators. Sterilized by gamma irradiation and supplied individually wrapped.
Disposable, color-coded cuvets are compatible with most electroporators.
  • Clear, medical grade polycarbonate for high visibility
  • Polished aluminum lowers arcing frequency
  • Round cap allows single-handed removal
  • FB102 has imbedded aluminum electrodes
Specifications
Sterility Sterile
Type Electroporation Cuvette
Capacity (Metric) 400 μL
Material Polycarbonate
Includes Imbedded aluminum electrodes
Width (Metric) Gap 2 mm
For Use With (Equipment) Electroporator