Learn More
Fisherbrand™ Cooling Block
Product Code. 16100381 Shop All Fisherbrand Traceable Products
Change view
Click to view available options
Capacity (Metric):
0.2 mL PCR tube strips
1 to 2 mL cryogenic vials
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Unit Size
16100381 1 to 2 mL cryogenic vials Each
17547026 0.2 mL PCR tube strips Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 16100381
Top Sellers. Mega Savings.
Use promo code 27997
View all promotions
Estimated Shipment: 08-12-2026
Add to basket
Add to basket
Fisherbrand Aluminum alloy cooling block keeps samples in place prior and during freezing process.
Specifications
Capacity (Metric) 1 to 2 mL cryogenic vials
Color Black
Material Aluminum
Shape Rectangular
For Use With (Application) null
Holds 30 (1 to 2 mL Cryogenic Vials)
Product Type Cryo Block
Quantity 1
Temperature Range (English) -122.8°F
Temperature Range (Metric) -86°C
Dimensions (L x W x H) 119 x 101 x 38 mm
Length (Metric) 16.3 cm
Width (Metric) 12.2 cm
Height (Metric) 4.3 cm