Learn More
Fisherbrand™ Cell Lifters
Product Code. 11577692 Shop All Fisherbrand Traceable Products
Change view
Click to view available options
Material:
Polyethylene
Unit Size:
Pack of 100
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Material Unit Size
11577692 Polyethylene Pack of 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11577692
Sample Preparation and Storage Essentials
Use promo code 27996
View all promotions
In Stock
Add to basket
Edge
Add to basket
Disposable lifters quickly remove cell layers
The Fisherbrand Cell Lifter is used with dishes and cell culture clusters for quick removal of cell layers. The one-piece design and chiseled blade provide improved control while minimizing damage during the cell removal process.
  • Sterilized and individually wrapped in peel-open packaging
  • Polyethylene copolymer blade with angled and chiseled edge
  • Maintains even contact with growth area in culture dishes
Specifications
Material Polyethylene
Sterility Sterile
For Use With (Application) Removal of cell layers
For Use With (Equipment) Dishes and cell culture clusters
Disposable Single-use
Length (English) Blade 0.7 in.
Length (English) Handle 7 in.
Length (Metric) Blade 2 cm
Length (Metric) Handle 17.78 cm
Edges Angled and Chiseled
Product Type Cell Lifter