Learn More
Fisherbrand™ Aluminum Weighing Dishes
Product Code. 11709418 Shop All Fisherbrand Products
Change view
Click to view available options
Capacity (Metric):
20 mL
42 mL
65 mL
70 mL
150 mL
Depth (Metric):
13 mm
16 mm
17 mm
17.3 mm
20 mm
Diameter (Metric) Inner Base:
44 mm
51 mm
68.2 mm
69 mm
96 mm
Shape:
Flanged Sides
Fluted Sides
Unit Size:
Pack of 100
Pack of 1000
Pack of 144
Pack of 8064
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Depth (Metric) Diameter (Metric) Inner Base Shape unitSize
11709418 70 mL 17.3 mm 68.2 mm Fluted Sides Pack of 1000
11799408 65 mL 16 mm 69 mm Fluted Sides Pack of 1000
11704069 42 mL 17 mm 51 mm Fluted Sides Pack of 8064
10297593 42 mL 17 mm 51 mm Fluted Sides Pack of 144
11789408 20 mL 13 mm 44 mm Fluted Sides Pack of 1000
11719418 150 mL 20 mm 96 mm Flanged Sides Pack of 1000
11518382 150 mL 20 mm 96 mm Flanged Sides Pack of 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Product Code. 11709418
Only null left
Add to basket
Add to basket
Aluminum dishes for weighing and dispensing
Choose Fisherbrand™ Aluminum Weight Dishes for weighing and dispensing.
  • With fluted sides
  • Lightweight, yet sturdy
  • Slightly sloping sides facilitate stacking
Specifications
Bottom Shape Flat
Capacity (Metric) 70 mL
Material Aluminum
Shape Fluted Sides
Type Weighing Dish
For Use With (Application) For weighing and dispensing
Depth (Metric) 17.3 mm
Diameter (Metric) Inner Base 68.2 mm
Diameter (Metric) Inner Top 72.6 mm
Diameter (Metric) 68.2 mm (Inner Base), 72.6 mm (Inner Top)