Learn More
Fisherbrand™ Adhesive Plate Seals
Product Code. 15963620 Shop All Fisherbrand Traceable Products
Change view
Click to view available options
Material:
Polypropylene
Unit Size:
Pack of 100
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Material Unit Size
15963620 Polypropylene Pack of 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15963620
  • ONLINE EXCLUSIVE PRICE
    483.18 EUR
    226.34 EUR / Pack of 100
    Save 256.84 EUR
    53% Off
Estimated Shipment: 04-12-2026
Add to basket
Add to basket
Seals are optically clear for PCR applications
  • Seals most plates
  • Rated from -40 to ±125°C
  • Polypropylene
Specifications
Color Clear
Material Polypropylene
Sterility Non-sterile
No. per Case 100
For Use With (Application) For PCR applications
Product Type Plate Seal
Temperature Range (Metric) -40°C to +125°C