Learn More
Fisherbrand™ Absorbent Underpads
GREENER_CHOICE
Product Code. 11917964 Shop All Fisherbrand Products
Change view
Click to view available options
Dimensions (L x W):
36 x 20 in.
Unit Size:
Pack of 125
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Dimensions (L x W) unitSize
11917964 36 x 20 in. Pack of 125
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11917964
Add to basket
Add to basket
High-absorbency fibers soak up spills.
  • Quickly absorb liquids and keep spills from spreading
  • Lie flat, hugging the benchtop to prevent sliding
  • Cushioned surface minimizes breakage of delicate labware
  • Polyethylene backing is chemical resistant
  • Spunbond top layer provides durability even when wet
  • Absorb up to 750mL/m2
TRUSTED_SUSTAINABILITY
Specifications
Product Type Absorbent Underpad
Material Polyethylene
Absorbancy 750 mL/m2
Length (English) 36 in.
Length (Metric) 910 mm
Width (English) 20 in.
Width (Metric) 510 mm
Dimensions (L x W) 36 x 20 in.
For Use With (Application) Quickly absorb liquids and keep spills from spreading