Learn More
Fisherbrand™ 96-Well Nonskirted PCR Plates
Product Code. 12104112 Shop All Fisherbrand Products
Change view
Click to view available options
Color:
Blue
Green
Natural
Red
Yellow
Unit Size:
Pack of 25
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color unitSize
12104112 Natural Pack of 25
12114112 Red Pack of 25
12124112 Blue Pack of 25
12144112 Yellow Pack of 25
12134112 Green Pack of 25
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Product Code. 12104112
Only null left
Add to basket
Add to basket
Compatible with most 0.2mL thermal cyclers and ABI PRISM™ 310, 3100, and 3700 capillary sequencers
  • Virgin polypropylene
  • Clear well bottom for sample visibility
  • Highly rigid plate deck
  • Raised rim design
  • Sealable using adhesive films and foils, heat seals, and cap strips
  • 300μL maximum volume when used with adhesive and heat seals
  • Cut-away corner and alphanumeric grid
  • Cleanroom produced
Specifications
Format Standard
Array 8 x 12
Color Natural
Product Type PCR Microplate
Skirt Style Non-skirt
Material Polypropylene
No. of Wells 96
For Use With (Application) PCR, RT-PCR applications
Rimmed Yes