Learn More
Fisherbrand™ 384-Well DeepWell Polypropylene Microplates
Shop All Fisherbrand Products
Click to view available options
Color:
Natural
Unit Size:
Pack of 60
Descrizione
- Excellent for storing DMSO and other strong chemicals and solvents
- Well shape maximizes sample retrieval
- Compatible with robotics and automation
- Rounded square wells minimize wicking (capillary action)
- Barcoding available by arrangement
- Sealable with chemical-resistant well cap mats
- Working volume range: 5 to 240μL/well
Specifica
Specifica
| Format | Standard |
| Array | 16 x 24 |
| Color | Natural |
| Product Type | 384-Well Microplate |
| Material | Polypropylene |
| Volume (Metric) Well | 252,000 μL |
| No. of Wells | 384 |
| Sterility | Non-sterile |
| Well Shape | Square |
| For Use With (Application) | For storage of compounds and nucleic acid libraries |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto