Learn More
Fisherbrand™ 3/8 in. Dot Labels
Product Code. 17837191 Shop All Fisherbrand Traceable Products
Change view
Click to view available options
Color:
Blue
Unit Size:
20 sheets
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Unit Size
17837191 Blue 20 sheets
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 17837191
Safety Starts Here: Save Up to 60%
Use promo code 28008
View all promotions
Estimated Shipment: 27-01-2027
Fisher branded Cryogenic Storage Labels use a special adhesive that is designed to withstand extreme cold temperatures as low as -196º C (-311º F). Our cryogenic storage labels can be printed or written on and will not bleed, smudge or smear. Engineered for you to use in your laser printer.
Specifications
Color Blue
Product Type Dot label laser sheet
Sterility Non-sterile
For Use With (Application) 0.5 to 1.5mL tubes
Quantity 192 dots/sheet; 20 sheets/Pk.
Diameter (English) 0.37 in.
Diameter (Metric) 9.55 mm