Learn More
Fisherbrand™ Glass Plates for FB-VE16-1 Vertical Electrophoresis System
Product Code. 11953045 Shop All Thermo Scientific Products
Change view
Click to view available options
Type:
Blank Glass Plate
Notched Glass Plate
Offset Glass Plate
Unit Size:
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Type Unit Size
11953045 Notched Glass Plate Each
11943035 Blank Glass Plate Each
11933035 Offset Glass Plate Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 11953045 Supplier Fisherbrand™ Supplier No. P114R
  • ONLINE EXCLUSIVE PRICE
    34.20 EUR
    32.30 EUR / Pack of 1
    Save 1.90 EUR
    5% Off
Estimated Shipment: 26-01-2027
Different styles of glass plates to choose from.
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) Owl P1-CST Multiple Gradient Gel Caster
Type Notched Glass Plate
Material Glass
Thickness (English) 0.12 in.
Thickness (Metric) 0.3 cm
Length (Metric) 16 cm
Width (Metric) 14 cm
Height (English) 0.12 in.
Height (Metric) 0.32 cm