Learn More
Fisherbrand™ Combs for FB-VE16-1 Vertical Electrophoresis System
Product Code. 11993045 Shop All Thermo Scientific Products
Change view
Click to view available options
Volume (Metric) Well:
51 μL
69 μL
96 μL
107 μL
129 μL
183 μL
201 μL
343 μL
Length (Metric) Footprint:
16 cm
20 cm
Teeth Count:
10
15
20
24
Width (Metric) Teeth:
0.29 cm
0.39 cm
0.61 cm
1.04 cm
Unit Size:
Each
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) Well Length (Metric) Footprint Teeth Count Width (Metric) Teeth Unit Size
11993045 129 μL 20 cm 20 0.39 cm Each
11973045 201 μL 20 cm 15 0.61 cm Each
11963045 107 μL 20 cm 15 0.61 cm Each
11913035 183 μL 20 cm 10 1.04 cm Each
11923035 343 μL 20 cm 10 1.04 cm Each
11983045 69 μL 20 cm 20 0.39 cm Each
11903055 51 μL 20 cm 24 0.29 cm Each
11913055 96 μL 16 cm 24 0.29 cm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Product Code. 11993045 Supplier Fisherbrand™ Supplier No. P120D
  • ONLINE EXCLUSIVE PRICE
    51.10 EUR
    49.00 EUR / Pack of 1
    Save 2.10 EUR
    4% Off
Estimated Shipment: 25-01-2027
Creates the wells in gels for electrophoresis
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) Owl P1-CST Multiple Gradient Gel Caster
Type Comb
Gel Dimensions 16 x 14 cm
Teeth Count 20
Thickness (Metric) Teeth 0.15 cm
Volume (Metric) Well 129 μL
Length (English) Footprint 7.87 in.
Length (English) Gel 6.3 in.
Length (Metric) Footprint 20 cm
Length (Metric) Gel 16 cm
Width (English) Gel 5.5 in.
Width (English) Teeth 0.15 in.
Width (Metric) Footprint 13 cm
Width (Metric) Gel 14 cm
Width (Metric) Teeth 0.39 cm
Height (English) Footprint 7.08 in.
Height (Metric) Footprint 18 cm