Learn More
Fisherbrand™ Traceable™ Handheld Digital Barometer
Product Code. 11724206 Shop All Fisherbrand Traceable Products
Change view
Click to view available options
Includes:
Battery
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Includes Unit Size
11724206 Battery Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11724206
  • ONLINE EXCLUSIVE PRICE
    343.00 EUR
    250.44 EUR / Pack of 1
    Save 92.56 EUR
    26% Off
 Restricted Item
Estimated Shipment: 30-12-2026
Add to basket
Add to basket
Keep track of your environment and your time with this multi-functional Traceable™ Handheld Digital Barometer.
  • Stopwatch/time of day clock
  • Barometric pressure trend displays current reading and previous 2, 4, 6, 8, 10 and 12 hours in a bar chart format
  • Minimum/maximum memory for altitudes reached during any time period
  • Barometer Accuracy: ±8 mbar (0.2363 in. Hg)
Certifications
Traceable to standards provided by NIST™. Individually serial-numbered Traceable Certificate is provided from an ISO 17025 calibration laboratory accredited by A2LA.
Specifications
Accuracy (Barometer) ±8 mbar
Resolution (Temperature) 0.1°C
Temperature Range (Metric) -10°C to +60°C
Temperature Range (English) 14°F to 140°F
Includes Battery
Product Type Handheld Digital Barometer