Learn More
Fisherbrand™ Traceable™ Dual-Display Conductivity Meter
Product Code. 11704226 Shop All Fisherbrand Traceable Products
Change view
Click to view available options
Display Type:
LCD
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Display Type Unit Size
11704226 LCD Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11704226
  • ONLINE EXCLUSIVE PRICE
    425.00 EUR
    357.53 EUR / Pack of 1
    Save 67.47 EUR
    15% Off
View all promotions
 Restricted Item
Estimated Shipment: 22-12-2026
Add to basket
Add to basket
Measures conductivity and temperature; includes NIST™ traceability certificate.
Large display
  • LCD is 1.375 in. high: simultaneously displays conductivity and temperature readings
  • Serial RS-232-C output connects to IBM-compatible computer or data logger
  • Optional Data Acquisition System DAS-3™ (11-661-22) or DAS-4™ Data Logger System (06-661-72) can capture, display, store and export data to an existing database
Measures conductivity in three ranges with NIST traceability
  • 0.0 to 199.9μmhos (0.1μmho resolution), 0.001 to 1.999mmhos (0.001mmho resolution), and 0.01 to 19.99 mohms (0.01 mmho resolution)
  • Accuracy is ±2% of full scale plus 1 digit
  • Automatic and manual temperature compensation from 0° to 60°C (32° to 140°F); resolution is 0.1° and accuracy is ±0.8°C
  • Certificate provided with each meter indicates instrument traceability to standards provided by the National Institute of Standards and Technology
  • Calibrated by an ISO 17025 calibration laboratory accredited by A2LA
Versatile Memory function
  • Recalls highest, lowest, and average readings at the touch of a button
  • HOLD button freezes display for later reading
  • Easy calibration: lets user adjust to a probe's K (constant) factor and to calibrate to solution standards
Compact and complete
  • Unit measures 1.25 W x 3 L x 7 in. H (3.2 x 7.6 x 17.8 cm)
  • Weighs 9.5 oz. (269 g)
Order Info
Accessories include Easy-Use™ 115 V adapter (11-661-23), DAS-3 Data Acquisition System (11-661-22), and DAS-4 Data Logger System (06-661-72) (all sold separately).
Certifications
Individually serial-numbered Traceable Certificate from an ISO 17025 calibration laboratory accredited by A2LA indicating instrument traceability to standards provided by NIST™.
Specifications
Includes Meter, Probe, 40 in. (101.6 cm) Cable, 9 V Alkaline Battery
Accuracy (Conductivity) ±2% FS +1 digit
Outputs Serial RS232C
For Use With (Application) Conductivity
Measurement Modes Conductivity, Temperature
Range (Conductivity) 0.0 to 199.9 Microohms
Temperature Compensation 0°C to 60°C
Temperature Range (Metric) 0 to 60°C
Resolution (Temperature) 0.1°C
Accuracy (Temperature) 0.8°C
Display Type LCD
Certifications/Compliance NIST
IP Rating None
Weight (Metric) 269 g
Weight (English) 9.5 oz.
Resolution 0.1°
Sensor Types Conductivity
Detection Range Multiple
Type Conductivity Meter
Accuracy ±2%
Length (English) 1.25 in.
Length (Metric) 3.2 cm
Width (English) 3 in.
Width (Metric) 7.6 cm
Height (English) 7 in.
Height (Metric) 17.8 cm
Battery Type Alkaline
Voltage 9 V