Learn More
Fisherbrand™ Stand and Clamp for Model 50 and 120 Sonic Dismembrator
Product Code. 12347338 Shop All Fisherbrand Traceable Products
Change view
Click to view available options
For Use With (Equipment):
Model 50 and 120 Sonic Dismembrator
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) Unit Size
12347338 Model 50 and 120 Sonic Dismembrator Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 12347338
  • ONLINE EXCLUSIVE PRICE
    121.00 EUR
    97.32 EUR / Pack of 1
    Save 23.68 EUR
    19% Off
Estimated Shipment: 19-11-2026
Add to basket
Add to basket
Stand and Converter clamp accessory for Model 50/120 Sonic Dismembrator. This accessory will properly support the converter and probe assembly during sonication.
This Fisher Scientific™ Stand is designed for use with Model 50 and Model 120 Sonic Dismembrators
  • Laboratory Support Stand with Converter clamp
  • For use with Model 50/120 Sonic Dismembrator
Specifications
Product Type Stand
Height (English) 24 in.
Height (Metric) 61 cm
For Use With (Equipment) Model 50 and 120 Sonic Dismembrator
Includes 0.5 in. dia. support rod and converter clamp