Learn More
Fisherbrand™ Sound Enclosure for Model 50 and 120 Sonic Dismembrator
Product Code. 12353745 Shop All Fisherbrand Traceable Products
Change view
Click to view available options
For Use With (Equipment):
Model 50/120 Sonic Dismembrator
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) Unit Size
12353745 Model 50/120 Sonic Dismembrator Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 12353745
  • ONLINE EXCLUSIVE PRICE
    1160.00 EUR
    848.09 EUR / Pack of 1
    Save 311.91 EUR
    26% Off
Estimated Shipment: 16-12-2026
Add to basket
Add to basket
Sound Enclosure for Model 50/120 Sonic Dismembrator. Sonic Dismembrators are extremely loud devices that can cause discomfort to the user and anyone nearby. The Sound Enclosure reduces noise by approximately 20 dBa and is made to work with all accessories. In addition to reducing noise, the Sound Enclosure has an internal support rod and converter mounting system. Any Fisherbrand probe or horn will be held safely and securely inside the unit.
This Fisher Scientific™ Sound Enclosure is designed for use with Model 50 and Model 120 Sonic Dismembrators
  • Sound Reducing Enclosure including stand and clamp
  • For use with Model 50/120 Sonic Dismembrator
  • Two ports are located on either side of the enclosure for coolant tubing or a temperature monitoring probe
  • Interior walls are lined with water-resistant acoustical foam
  • Door has a window so experiments can be visually monitored
  • Dimension: 50.8 x 25.4 x 25.4cm (20 x 10 x 10 in.)
Specifications
Product Type Sound Enclosure
Dimensions (L x W x H) 12 x 12 x 20 in.
Height (English) 20 in.
For Use With (Application) Ideal for sound reduction
For Use With (Equipment) Model 50/120 Sonic Dismembrator
Includes Internal support rod and converter mounting system