Learn More
Fisherbrand™ Probes for Model 50 and 120 Sonic Dismembrator
Product Code. 12921181 Shop All Fisherbrand Traceable Products
Change view
Click to view available options
Volume Range (Metric):
0.2 to 5 mL
0.5 to 15 mL
10 to 50 mL
Diameter (Metric):
2 mm
0.3 cm
0.63 cm
Amplitude:
120 μm maximum
180 μm maximum
200 μm maximum
Unit Size:
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume Range (Metric) Diameter (Metric) Amplitude Unit Size
12921181 0.2 to 5 mL 2 mm 200 μm maximum Each
12911181 0.5 to 15 mL 0.3 cm 180 μm maximum Each
12931181 10 to 50 mL 0.63 cm 120 μm maximum Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 12921181
  • ONLINE EXCLUSIVE PRICE
    338.00 EUR
    272.49 EUR / Pack of 1
    Save 65.51 EUR
    19% Off
Estimated Shipment: 07-10-2026
Add to basket
Add to basket
Image shows (left to right): 4423, 4422, 4435.
These Fisher Scientific™ Probes are designed for use with Model 50 and Model 120 Sonic Dismembrators.
  • 1/8 in. Probe microtip for processing 0.5 to 15mL volumes
  • 5/64 in. Probe microtip for processing 0.2 to 5mL volumes
Specifications
Amplitude 200 μm maximum
Product Type Probe
Volume Range (Metric) 0.2 to 5 mL
Diameter (English) 0.078 in. (5/64 in.)
Diameter (English) Tip 5/64 in.
Diameter (Metric) 2 mm
Diameter (Metric) Tip 2 mm
For Use With (Equipment) Model 50 and 120 sonic dismembrator
Packaging Quantity 1 probe