Learn More
Fisherbrand™ Pocket-Sized Scientific Calculator
Product Code. 11957586 Shop All Fisherbrand Traceable Products
Change view
Click to view available options
Display Type:
Digital
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Display Type Unit Size
11957586 Digital Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11957586
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Includes
Backup battery and instructions
Ideal for virtually all routine calculations performed in scientific labs, including square roots, percentages, and raising to powers
Performs more than 50 functions including science, math, common/hyperbolic trigonometric, logs/natural logs, powers, roots, reciprocals, factorials, percentages, hexadecimals, random number generation, and degree/decimal conversion.
  • Statistical features include standard deviation, mean, sum of squares, sum of data and much more
  • Easy-to-use, raised keys and large, easy-to-read, 0.25 in.H (0.6cm), 10-digit, two-digit exponent LCD
  • Three-key memory and “Forever Memory” store data when the unit is off
  • Automatic power-down function conserves battery life
Order Info
Dimensions: 5L x 3W x 0.5 in.H (12.7 x 7.6 x 1.3cm). Net weight: 1.5 oz. (43g)
Specifications
Product Type Scientific Calculator (Pocket-sized)
Display Type Digital
Key Functions Science, math, common/hyperbolic trigonometric, logs/natural logs, powers, roots, reciprocals, factorials, percentages, hexadecimals, random number generation and degree/decimal conversion
Keypad Raised keys
Memory 3-key
Dimensions (L x W) 5 .0 x 3.0 in.
Height (English) 0.5 in.
Height (Metric) 1.3 cm
Length (English) 5 in.
Length (Metric) 12.7 cm
Width (English) 3 in.
Width (Metric) 7.6 cm
Weight (English) 1.5 oz.
Weight (Metric) 42.5 g
For Use With (Application) Over 50 scientific functions
Includes Backup battery and instructions