Learn More
Fisherbrand™ Isotemp™ Gable Covers for Water Baths
Product Code. 15167843 Shop All Fisherbrand Products
Change view
Click to view available options
For Use With (Equipment):
Model 202
Model 220, 228
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) unitSize
15167843 Model 202 Each
15197843 Model 220, 228 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15167843 Supplier Fisherbrand™ Supplier No. 1546232Q
Only null left
Add to basket
Add to basket
Reduce moisture loss and water accumulation on the bench.
Stainless steel gable covers for water baths. Reduce moisture loss and water accumulation on the bench
  • 1 year warranty
  • Material: Stainless steel
Warranty
One year
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) Model 202
Material Stainless Steel
Warranty 1 Year