Learn More
Fisherbrand™ Filling Solutions for accumet™ ISE Electrodes, Fluoride
Product Code. 12952920 Shop All Fisherbrand Traceable Products
Change view
Click to view available options
For Use With (Equipment):
Accumet Chloride ISE Electrode
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) Unit Size
12952920 Accumet Chloride ISE Electrode Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 12952920
  • ONLINE EXCLUSIVE PRICE
    111.00 EUR
    81.53 EUR / Pack of 1
    Save 29.47 EUR
    26% Off
 Restricted Item
Estimated Shipment: 23-11-2026
For use with accumet™ ISE Electrodes.
Five 60mL bottles
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) Accumet Chloride ISE Electrode
Type Filling Solution
Filling Solution Type Chloride
Includes Five 60mL bottles