Learn More
Fisherbrand™ Circular Bottom Desi-Vac™ Container Desiccators with Vacuum Pump
Product Code. 11744366 Shop All Fisherbrand Products
Change view
Click to view available options
Volume (Metric):
700 mL
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) unitSize
11744366 700 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Do not autoclave or expose container to organic solvents or their vapors. Do not use concentrated acids as desiccants.
Product Code. 11744366
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Do not autoclave or expose container to organic solvents or their vapors. Do not use concentrated acids as desiccants.
Desiccator with built-in manual vacuum pump eliminates need for separate pump, tubing and stopcock
Desiccator with built-in manual vacuum pump eliminates need for separate pump, tubing and stopcock
  • Ideal for drying, preserving, storing and shipping samples
  • Self-contained unit holds a vacuum of 14.7 in.Hg (373mm) in excess of 24 hours
  • Rubber O-ring in cover ensures a tight seal
  • To remove cover, simply push release button to allow air into container
  • Clear polycarbonate body, metal pump handle, chemical-resistant ABS plastic pump and cover
  • No plate required
Ideal for drying, preserving, storing and shipping samples
Order Info
Available in volumes of 600, 850, or 1100mL. Can be used with Fisherbrand™ Humidity Sponge™ Desiccant.
Specifications
Bottom Shape Circular
Description 11.43 x 11.43 x 12.7cm ((4.5 x 4.5 x 5.0 in.)
Material Polycarbonate
Product Type Desiccator
Includes Plastic pump handle, chemical-resistant ABS plastic vacuum pump and cover, release button on top, O-Ring-sealed cover
Volume (English) 42 cu. in.
Volume (Metric) 700 mL
Width (English) 4.5 in.
Width (Metric) 11.43 cm
Height (Metric) 12.7 cm
Height (English) 5 in.
Depth (English) 5.0 in.
Depth (Metric) 12.7 cm