Learn More
Fisherbrand™ accumet™ Automatic Temperature Compensation Probes - Mercury - Free
Product Code. 11530234 Shop All Fisherbrand Products
Change view
Click to view available options
Connector Types:
0.25 in. Phone
Mini-Phone
Material:
316 Stainless Steel
Stainless Steel
For Use With (Equipment):
Accumet benchtop and portable meter
Accumet model AP63, AP62, AP61
accumet AB, AR, XL, Basic, benchtop meters
Unit Size:
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Connector Types Material For Use With (Equipment) unitSize
11530234 0.25 in. Phone Stainless Steel Accumet benchtop and portable meter Each
10236064 Mini-Phone 316 Stainless Steel accumet AB, AR, XL, Basic, benchtop meters Each
11540234 Mini-Phone Stainless Steel Accumet model AP63, AP62, AP61 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 11530234
  • ONLINE EXCLUSIVE PRICE
    412.00 EUR
    305.95 EUR / Pack of 1
    Save 106.05 EUR
    25% Off
Estimated Shipment: 22-09-2026
Add to basket
Add to basket
For accumet™ benchtop meters.
For use with accumet Meters
  • Signal solution temperature to meter for automatic compensation
  • Permit temperature readout with meters having this feature
  • Probes for accumet AP Series portable meters have waterproof connector
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) Accumet benchtop and portable meter
Material Stainless Steel
Type ATC
Connector Types 0.25 in. Phone
Length (Metric) Cable 0.9 m