Learn More
Fisher BioReagents™ Microbiology Media Additives: Yeast Extract
Product Code. 10225203
Change view
Click to view available options
Quantity:
100 g
500 g
2000 g
Unit Size:
100g
2kg
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10225203 500 g 500g
10255153 100 g 100g
10215203 2000 g 2kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 10225203 Supplier Fisher BioReagents™ Supplier No. BP1422500
 Restricted Item
Estimated Shipment: 12-11-2026
Add to basket
Additive for preparing bacterial nutrient media
Water soluble portion of autolyzed fresh yeast used in bacteriological culture media
TRUSTED_SUSTAINABILITY
Specifications
Description Bacterial Culture
Form Dehydrated
Grade Molecular genetics
Media Type Yeast Extract
Product Type Microbiology Media
Packaging Type Poly Bottle
Quantity 500 g