Learn More
Plum™ First Aid Station
Product Code. 15741147
Change view
Click to view available options
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. unitSize
15741147 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15741147 Supplier Plum™ Supplier No. P5101
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
For easy access to first aid for small accidents. All-In-One solution ideal for almost any workspace.
  • First aid station with a 500 mL Plum Eye Wash bottle, a QuickFix plaster dispenser with 45 elastic and 45 waterproof plasters, and a QuickClean wound cleanser dispenser with 40 wound cleansing wipes
  • 0.9 % sterile sodium chloride – similar to that of the human eye. Provides quick relief if dust or dirt comes into contact with an eye or dry eye conditions
  • Handy and mobile access to CE labelled eyewash - with a 3-year expiry date
  • Immidiate access to waterproof and elastic plaster. Each CE-labelled plaster is individually packed and can be used with one hand
  • QuickClean napkins for easy cleansing of minor injuries and scratches
  • Specifications
    For Use With (Application) All-In-One solution ideal for almost any workspace
    Color Green
    Width (Metric) 515 mm
    Height (Metric) 300 mm