missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FIPI, MedChemExpress
Description
FIPI is a derivative of halopemide which potently inhibits both PLD1 and PLD2 with IC50s of 25 nM and 20 nM, respectively.
Specifications
Specifications
| Chemical Name or Material | FIPI |
| CAS | 939055-18-2 |
| Color | Earth Yellow |
| Purity Grade Notes | Research |
| Molecular Formula | C23H24FN5O2 |
| Quantity | 5 mg |
| Synonym | 5-Fluoro-2-indolyl deschlorohalopemide |
| Solubility Information | DMSO : 14.67 mg/mL (34.81 mM; Need ultrasonic and warming) |
| SMILES | O=C(C(N1)=CC2=C1C=CC(F)=C2)NCCN3CCC(N4C5=CC=CC=C5NC4=O)CC3 |
| Molecular Weight (g/mol) | 421.47 |
| Show More |
Research purposes only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?