Learn More
Thermo Scientific™ Finntip™ Stepper Pipette Tips
GREENER_CHOICE
Product Code. 10626111
Change view
Click to view available options
Volume (Metric):
500 μL
1250 μL
2.5 mL
5 mL
12.5 mL
25 mL
50 mL
Sterility:
Non-sterile
Sterile
Unit Size:
Pack of 10
Pack of 100
Pack of 20
Pack of 25
Pack of 50
14 product options available for selection
Product selection table with 14 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) Sterility unitSize
10626111 500 μL Non-sterile Pack of 100
10411891 500 μL Sterile Pack of 50
10185900 1250 μL Non-sterile Pack of 100
11782574 1250 μL Sterile Pack of 50
10542481 2.5 mL Non-sterile Pack of 100
11792574 2.5 mL Sterile Pack of 50
10762111 5 mL Non-sterile Pack of 50
11702584 5 mL Sterile Pack of 25
11742574 12.5 mL Non-sterile Pack of 50
11712584 12.5 mL Sterile Pack of 25
10401891 25 mL Non-sterile Pack of 20
11722584 25 mL Sterile Pack of 10
10083630 50 mL Non-sterile Pack of 10
10750981 50 mL Sterile Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 14 options available.
14 options
Product Code. 10626111 Supplier Thermo Scientific™ Supplier No. 9404170
  • ONLINE EXCLUSIVE PRICE
    196.00 EUR
    104.76 EUR / Pack of 100
    Save 91.24 EUR
    46% Off
View all promotions
In Stock
Add to basket
Add to basket
Integrate Thermo ScientificTM FinntipTM Stepper Pipette Tips with the Thermo ScientificTM FinnpipetteTM Stepper Pipetter.
Integrate Thermo Scientific™ Finntip™ Stepper Pipette Tips with the Thermo Scientific™ Finnpipette™ Stepper Pipetter.
  • Nonsterile tips are bulk packaged; sterile tips are individually wrapped
TRUSTED_SUSTAINABILITY
Specifications
Volume (Metric) 500 μL
Volume (Metric) Dispensing 10, 20, 30, 40, 50 μL
Filtered Non-filtered
Sterility Non-sterile
Certifications/Compliance ISO 9001, ISO 14001
Format Bulk
For Use With (Equipment) Thermo Scientific FinnPipette Stepper
Product Line FinnTip