Learn More
Thermo Scientific™ Finnpipette™ Stands
Click to view available options
Product Type:
Carousel Stand Adapter
Carousel Stand with Cover
Pipette Stand
Shelf Hanger
Holds:
1
3
Electronic Pipette
Manual Pipette
Manual Pipettes (3)
Manual Pipettes (6)
Manual Pipettes (6) or Electronic Pipettes (3)
Multi-channel Manual Pipette
Packungsgröße:
1 set
Each
Beschreibung
Provide safe and convenient storage for manual and electronic pipettes using Thermo Scientific™ Finnpipette™ Stands. Available options include options for single stand, F-stand, multi-stand, multichannel and carousel.
Finnpipette F-Stand
- Accommodates six pipettes
- Sturdy crossbar structure with slot for each pipette for secure storage
- Suitable for both single channel and multichannel models
Finnpipette Multichannel Stand
- Multi-position stand
- Can hold any multichannel Finnpipette
- Pipette can be stored in upright or horizontal position
- Small footprint
Finnpipette Carousel Stand
- Rotating, portable
- Transparent lid protects pipettes from contamination
- Shipped with three adapters for electronic Finnpipette Novus pipettes and five adapters for manual Finnpipette pipettes
Finnpipette Novus Stand
- For convenient storage of electronic Finnpipette Novus pipette
- Accommodates both single and multichannel Finnpipette Novus models with ease
Spezifikation
Spezifikation
| Product Type | Pipette Stand |
| Description | Pipette Mini-Stand, gray |
| Color | Gray |
| Holds | Manual Pipettes (3) |
| Positions | 3 |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur