Learn More
Thermo Scientific™ Finger Tite Connectors
Product Code. 10584465 Shop All Thermo Scientific Products
Change view
Click to view available options
Type:
Female Nut
Split/Splitless and FID Starter Kit
Split/Splitless and MS Starter Kit
Unit Size:
Each
Pack of 5
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Type unitSize
10584465 Female Nut Pack of 5
10177804 Split/Splitless and MS Starter Kit Each
10321485 Split/Splitless and FID Starter Kit Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 10584465 Supplier Thermo Scientific™ Supplier No. 290ST130
Only null left
Add to basket
Add to basket
Ensure reliable, leak-free connections for optimal GC performance using these Finger Tite Connectors.
Ensure reliable, leak-free connections for optimal GC performance using Thermo Scientific™ Finger Tite Connectors. The perfect metal-to-metal seal eliminates contamination and produces lower air/water background noise compared to conventional graphite/polymer ferrules. They are easy to handle even with a hot injector/detector and do not require the use of tools. Unlike other ferrules, they do not require retightening after installation.
Recommended Use
  • Thermo Scientific™ and Agilent™ GC instruments
  • Connectors for SSL and MS systems
TRUSTED_SUSTAINABILITY
Specifications
Type Female Nut
Description Female Nut
For Use With (Equipment) TRACE GC Ultra and FOCUS GC Instruments