Learn More
Fimasartan, TRC
Product Code. 30403537
Change view
Click to view available options
Quantity:
10 mg
100 mg
250 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30403537 10 mg
30391362 100 mg
30391027 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30403537 Supplier Toronto Research Chemicals Supplier No. TRCF34204010MG
 Restricted Item
Estimated Shipment: 14-08-2026
Add to basket
Add to basket
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 247257-48-3
Molecular Formula C27 H31 N7 O S
Molecular Weight (g/mol) 501.65
Synonym 5-Pyrimidineethanethioamide, 2-butyl-1,6-dihydro-N,N,4-trimethyl-6-oxo-1-[[2'-(2H-tetrazol-5-yl)[1,1'-biphenyl]-4-yl]methyl]-, 5-Pyrimidineethanethioamide, 2-butyl-1,6-dihydro-N,N,4-trimethyl-6-oxo-1-[[2'-(1H-tetrazol-5-yl)[1,1'-biphenyl]-4-yl]methyl]- (9CI), 2-Butyl-1,6-dihydro-N,N,4-trimethyl-6-oxo-1-[[2'-(2H-tetrazol-5-yl)[1,1'-biphenyl]-4-yl]methyl]-5-pyrimidineethanethioamide, 2-n-Butyl-5-dimethylaminothiocarbonylmethyl-6-methyl-3-[[2'-(1H-tetrazol-5-yl)biphenyl-4-yl]methyl]pyrimidin-4(3H)-one, Fimasartan, Kanarb
IUPAC Name 2-[2-butyl-4-methyl-6-oxo-1-[[4-[2-(1H-tetrazol-5-yl)phenyl]phenyl]methyl]pyrimidin-5-yl]-N,N-dimethylethanethioamide
SMILES CCCCC1=NC(=C(CC(=S)N(C)C)C(=O)N1Cc2ccc(cc2)c3ccccc3c4nnn[nH]4)C
Specifications
Quantity 10 mg
InChI Formula InChI=1S/C27H31N7OS/c1-5-6-11-24-28-18(2)23(16-25(36)33(3)4)27(35)34(24)17-19-12-14-20(15-13-19)21-9-7-8-10-22(21)26-29-31-32-30-26/h7-10,12-15H,5-6,11,16-17H2,1-4H3,(H,29,30,31,32)
Formula Weight 501.2311
Chemical Name or Material Fimasartan