Learn More
Thermo Scientific™ Fiberlite™ F21-8 x 50y Fixed-Angle Rotor
GREENER_CHOICE
Product Code. 12141680
Change view
Click to view available options
For Use With (Equipment):
Thermo Scientific Sorvall RC 6 Plus Centrifuge
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) unitSize
12141680 Thermo Scientific Sorvall RC 6 Plus Centrifuge Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 12141680 Supplier Thermo Scientific™ Supplier No. 46923
Separate samples safely at maximal g-force using the fixed angle rotor with bioseal technology.
Safely separate samples at maximal g-force using the Thermo Scientific™ Fiberlite™ F21-8 x 50y Fixed-Angle Rotor with Bioseal technology.
Features:
Fiberlite Rotors:
  • Reduce run times with higher G-forces achieved with lightweight carbon fiber rotors
  • Ensure structural integrity with corrosion- and fatigue-resistant design
  • Backed by a 15-year warranty
Compatible with:
Thermo Scientific Sorvall™ RC-6 Plus Series, Thermo Scientific™ Jouan KR22i centrifuge, Beckman™ J2 Series, Thermo Scientific™ Nalgene™ bottles and tubes
Recommended for:
Subcellular fractions, bacteria pelleting, lysate clarification, cell membranes, sucrose gradient isolation of mitochondria, endoplasmic reticulum and gradient separation of cells with Percoll, Ficoll, Optiprep or Nycodenz solutions
Note:
†Performance may vary by centrifuge model.
TRUSTED_SUSTAINABILITY
Specifications
Description Fiberlite F21-8 x 50y Fixed-Angle Rotor
Material Carbon Fiber Composite
Type Fixed Angle Rotor
Capacity 8 x 50 mL
Max. RCF 52,400 x g
Max. Speed 21,000 rpm
Radius (Metric) 106 mm
Radius (Metric) Min. 33 mm
K Factor 669
Max. Tube Length 115 mm
Max. Tube Diameter 29 mm
Net Weight 9.9 lb. (4.49 kg)
For Use With (Equipment) Thermo Scientific Sorvall RC 6 Plus Centrifuge
Angle 34°
Min. RCF 16,299 x g
Biocontainment Yes