Learn More
Brady™ Fiberglass-Reinforced Nylon Miniature Locking System for Circuit Breakers
Product Code. 15603517
Change view
Click to view available options
Quantity:
1 Ea.
6 Pk.
Description:
Pin-In Standard
Pin-Out Standard
Unit Size:
Each
Pack of 6
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Description unitSize
15603517 6 Pk. Pin-In Standard Pack of 6
11751945 1 Ea. Pin-Out Standard Each
15693507 6 Pk. Pin-Out Standard Pack of 6
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 15603517 Supplier Brady™ Supplier No. 090848
In Stock
Add to basket
Add to basket
A safe and effective method for locking out European Circuit Breakers.
  • Requires no tools to install
  • Lockout easily installs with the aid of a push button
  • Tie-bar style lockout has a thumb wheel for quick application
Specifications
Product Type Miniature Locking System for Circuit Breakers
Alternate Part Number ELEC. CIRCUIT BREAKER L/O EURO PIS PK6
Color Red/Black
Material Fiberglass-reinforced Nylon
Description Pin-In Standard
Diameter (Metric) 24.13 mm
Dimensions (D x W x H) 13.72 x 24.13 x 41.7 mm
Lockout Type Pin In
Quantity 6 Pk.
Width (Metric) 24.13 mm