missing translation for 'onlineSavingsMsg'
Learn More

Fertilysin, MP Biomedicals™

Product Code. 11455481 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11455481 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11455481 Supplier MP Biomedicals Supplier No. 0215805091
  • ONLINE EXCLUSIVE PRICE
    381.00 EUR
    362.25 EUR / 1g
    Save 18.75 EUR
    4% Off
 Restricted Item
Estimated Shipment: 05-02-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

White to off-white powder used for biochemical research.

Specifications

CAS 1477-57-2
Molecular Formula C12H20Cl4N2O2
Quantity 1 g
Synonym fertilysin, bis-diamine, n,n'-octane-1,8-diyl bis 2,2-dichloroacetamide, fertilysine, n,n'-octamethylenebis dichloroacetamide, bisdiamine, fertilysine n, n,n'-bis dichloroacetyl-1,8-octamethylenediamine, n,n'-octamethylenebis 2,2-dichloroacetamide, n,n'-bis dichloracetyl-1,8-octanediamine
InChI Key FAOMZVDZARKPFJ-UHFFFAOYSA-N
SMILES C(CCCCNC(=O)C(Cl)Cl)CCCNC(=O)C(Cl)Cl
IUPAC Name 2,2-dichloro-N-[8-[(2,2-dichloroacetyl)amino]octyl]acetamide
Molecular Weight (g/mol) 366.104
PubChem CID 15134
Formula Weight 366.1
Percent Purity 95%
Physical Form Solid
Chemical Name or Material N, N'-Octamethylen-bis(2, 2-Dichloroacetamide)
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.