Learn More
Ferrocene, Sigma-Aldrich™
Product Code. 15874141
Change view
Click to view available options
Quantity:
50 g
Unit Size:
50g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15874141 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
MQ200 Graded - For Research and Non-regulated Industrial Applications
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 15874141 Supplier Sigma Aldrich Supplier No. 8039780050
 Restricted Item
Estimated Shipment: 18-11-2026
MQ200 Graded - For Research and Non-regulated Industrial Applications
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
For synthesis
TRUSTED_SUSTAINABILITY
Specifications
CAS 102-54-5
Density 1.49 g/cm3 at 20°C
Molecular Formula (C5H5)2Fe
MDL Number MFCD00001427
Quantity 50 g
Synonym Ferrocene, Dicyclopentadienyliron, Iron dicyclopentadienyl, Ferrocene
Molecular Weight (g/mol) 186.03
Grade Research