Learn More
Thermo Scientific™ Sodium Acetate Solution (3 M), pH 5.2
Product Code. 10190890 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
5 x 1 mL
Unit Size:
Pack of 5
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10190890 5 x 1 mL Pack of 5
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10190890 Supplier Thermo Scientific™ Supplier No. R1181
  • ONLINE EXCLUSIVE PRICE
    28.52 EUR
    26.65 EUR / Pack of 5
    Save 1.87 EUR
    6% Off
View all promotions
 Restricted Item
In Stock
Add to basket
Edge
Add to basket
Precipitate DNA and RNA with this 0.22μm membrane-filtered, sterile, nuclease-free solution of sodium acetate.
3 M Sodium Acetate Solution, pH 5.2 is a 0.22 μm membrane filtered, sterile, nuclease-free solution of sodium acetate. This solution is routinely used in DNA and RNA precipitation.
Routinely used in DNA and RNA precipitation.
Recommended for:
DNA precipitation; RNA precipitation.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Sodium Acetate
Product Type Sodium Acetate Solution
Quantity 5 x 1 mL
Form Liquid