Learn More
Hellma FEP Tubing Set
Product Code. 15776563
Change view
Click to view available options
Diameter (Metric) Inner:
1.1 mm
Unit Size:
1 set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Diameter (Metric) Inner Unit Size
15776563 1.1 mm 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15776563 Supplier Hellma Supplier No. 040111722
  • ONLINE EXCLUSIVE PRICE
    49.65 EUR
    47.94 EUR / 1 sets
    Save 1.71 EUR
    3% Off
Estimated Shipment: 16-03-2027
Suitable for use with compact and 3-in-1, flow-through cells with one short and one long screw fitting.
  • Fluorinated ethylene propylene (FEP) construction
  • FEP is the preferred material for making small diameter tubing of continuous lengths
  • Chemical and dielectric properties similar to PTFE
  • One short screw fitting
  • One long screw fitting
  • For compact and 3-in-1 cells
Specifications
Description Length: 500 mm
Diameter (Metric) Inner 1.1 mm
Material FEP
Diameter (Metric) Outer 1.9 mm
Includes One short screw connection, one long screw connection