missing translation for 'onlineSavingsMsg'
Learn More

Fenpropidin, TRC

Product Code. 30398295
Change view
Click to view available options
Quantity:
50 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30398295 50 mg
30393098 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30398295 Supplier Toronto Research Chemicals Supplier No. TRCF24947050MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 67306-00-7
Molecular Formula C19 H31 N
Molecular Weight (g/mol) 273.46
Synonym 1-[3-[4-(1,1-Dimethylethyl)phenyl]-2-methylpropyl]piperidine, F 3240, Fenpropidin, Fenpropidine, Mildin, Patrol, Piperidine, 1-[3-[4-(1,1-dimethylethyl)phenyl]-2-methylpropyl]-
IUPAC Name 1-[3-(4-tert-butylphenyl)-2-methylpropyl]piperidine
SMILES CC(CN1CCCCC1)Cc2ccc(cc2)C(C)(C)C

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C19H31N/c1-16(15-20-12-6-5-7-13-20)14-17-8-10-18(11-9-17)19(2,3)4/h8-11,16H,5-7,12-15H2,1-4H3
Formula Weight 273.2457
Percent Purity >95%
Chemical Name or Material Fenpropidin
Grade GC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.