Learn More
Fenoprofen Calcium Dihydrate, LoGiCal
Product Code. 30462495
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30462495 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30462495 Supplier LoGiCal Supplier No. LGCFOR0790.00
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
LoGiCal's certified reference materials as neats, native solutions and stable isotope labelled solutions, with expert documentation and support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Fenoprofen calcium dihydrate
CAS 71720-56-4
Analyte or Component Names Fenoprofen calcium dihydrate
Shipping Condition Room Temperature
Molecular Formula 2 C15 H13 O3 . Ca . 2 H2 O
Certifications/Compliance ISO 9001
Synonym Fenoprofen Calcium Dihydrate, Fenoprofen calcium salt dihydrate, Benzeneacetic acid, alpha-methyl-3-phenoxy-, calcium salt, hydrate (2:1:2), Benzeneacetic acid, alpha-methyl-3-phenoxy-, calcium salt, dihydrate (9CI)
InChI Formula InChI=1S/2C15H14O3.Ca.2H2O/c2*1-11(15(16)17)12-6-5-9-14(10-12)18-13-7-3-2-4-8-13;;;/h2*2-11H,1H3,(H,16,17);;2*1H2/q;;+2;;/p-2
Solution Type Neat
SMILES O.O.[Ca+2].CC(C(=O)[O-])c1cccc(Oc2ccccc2)c1.CC(C(=O)[O-])c3cccc(Oc4ccccc4)c3
IUPAC Name calcium;2-(3-phenoxyphenyl)propanoate;dihydrate
Industry Type Clinical Testing
Molecular Weight (g/mol) 558.63
Quantity 10 mg
Formula Weight 558.1567 g/mol