missing translation for 'onlineSavingsMsg'
Learn More

Fenofibrate, TRC

Product Code. 30135200
Change view
Click to view available options
Quantity:
100 mg
1 g
25 g
Unit Size:
100mg
1g
25g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30135200 25 g 25g
30134127 1 g 1g
17943515 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30135200 Supplier Toronto Research Chemicals Supplier No. TRCF24864025G
 Restricted Item
Estimated Shipment: 27-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 49562-28-9
Molecular Formula C20 H21 Cl O4
Molecular Weight (g/mol) 360.83
Synonym Fenofibrate, 2-[4-(4-Chlorobenzoyl)phenoxy]-2-methylpropanoic acid 1-methylethyl ester, Ankebin, Antara, Clorofibrate, Elasterin, Fenobrate, Fenogal, Fenotard, Isopropyl 2-[p-(p-chlorobenzoyl)phenoxy]-2-methylpropionate, LF 178, Lipanthyl, Lipantil, Lipidil, Lipidil Supra, Lipirex, Lipoclar, Lipofene, Liposit, Lipsin, NSC 281319, Nolipax, Procetofen, Procetofene, Procetoken, Protolipan, Secalip, TriCor
IUPAC Name propan-2-yl 2-[4-(4-chlorobenzoyl)phenoxy]-2-methylpropanoate
SMILES CC(C)OC(=O)C(C)(C)Oc1ccc(cc1)C(=O)c2ccc(Cl)cc2

Specifications

Quantity 25 g
InChI Formula InChI=1S/C20H21ClO4/c1-13(2)24-19(23)20(3,4)25-17-11-7-15(8-12-17)18(22)14-5-9-16(21)10-6-14/h5-13H,1-4H3
Formula Weight 360.1128
Percent Purity >95%
Chemical Name or Material Fenofibrate
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.