Learn More
Fenclofenac, TRC
Product Code. 30392072
Change view
Click to view available options
Quantity:
50 mg
250 mg
500 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30392072 50 mg
30402753 250 mg
30406155 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30392072 Supplier Toronto Research Chemicals Supplier No. TRCF24750550MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 34645-84-6
Molecular Formula C14 H10 Cl2 O3
Molecular Weight (g/mol) 297.13
Synonym Benzeneacetic acid, 2-(2,4-dichlorophenoxy)-, Fenclofenac, Flenac, R 67408, RX 67408, [o-(2,4-Dichlorophenoxy)phenyl]acetic acid
IUPAC Name 2-[2-(2,4-dichlorophenoxy)phenyl]acetic acid
SMILES OC(=O)Cc1ccccc1Oc2ccc(Cl)cc2Cl
Specifications
Quantity 50 mg
InChI Formula InChI=1S/C14H10Cl2O3/c15-10-5-6-13(11(16)8-10)19-12-4-2-1-3-9(12)7-14(17)18/h1-6,8H,7H2,(H,17,18)
Formula Weight 296.0007
Percent Purity >95%
Chemical Name or Material Fenclofenac
Grade HPLC