Learn More
Fenclofenac, Mikromol™
Product Code. 30051833
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30051833 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30051833 Supplier Mikromol Supplier No. MM3075.00
 Restricted Item
Estimated Shipment: 17-08-2026
Add to basket
Add to basket
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
Fenclofenac
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Fenclofenac
CAS 34645-84-6
Analyte or Component Names Fenclofenac
Shipping Condition Room Temperature
Molecular Formula C14 H10 Cl2 O3
Certifications/Compliance ISO 17025
Synonym Benzeneacetic acid, 2-(2,4-dichlorophenoxy)-, Fenclofenac, Flenac, R 67408, RX 67408, [o-(2,4-Dichlorophenoxy)phenyl]acetic acid
InChI Formula InChI=1S/C14H10Cl2O3/c15-10-5-6-13(11(16)8-10)19-12-4-2-1-3-9(12)7-14(17)18/h1-6,8H,7H2,(H,17,18)
SMILES OC(=O)Cc1ccccc1Oc2ccc(Cl)cc2Cl
IUPAC Name 2-[2-(2,4-dichlorophenoxy)phenyl]acetic acid
Molecular Weight (g/mol) 297.13
Quantity 100 mg
Formula Weight 296.0007 g/mol