Learn More
Fenclofenac, LoGiCal
Product Code. 30458357
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30458357 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30458357 Supplier LoGiCal Supplier No. LGCFOR3075.00
 Restricted Item
Estimated Shipment: 17-08-2026
Add to basket
Add to basket
LoGiCal's certified reference materials as neats, native solutions and stable isotope labelled solutions, with expert documentation and support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Fenclofenac
CAS 34645-84-6
Analyte or Component Names Fenclofenac
Shipping Condition Room Temperature
Molecular Formula C14 H10 Cl2 O3
Certifications/Compliance ISO 9001
Synonym Benzeneacetic acid, 2-(2,4-dichlorophenoxy)-, Fenclofenac, Flenac, R 67408, RX 67408, [o-(2,4-Dichlorophenoxy)phenyl]acetic acid
InChI Formula InChI=1S/C14H10Cl2O3/c15-10-5-6-13(11(16)8-10)19-12-4-2-1-3-9(12)7-14(17)18/h1-6,8H,7H2,(H,17,18)
Solution Type Neat
SMILES OC(=O)Cc1ccccc1Oc2ccc(Cl)cc2Cl
IUPAC Name 2-[2-(2,4-dichlorophenoxy)phenyl]acetic acid
Industry Type Clinical Testing
Molecular Weight (g/mol) 297.13
Quantity 10 mg
Formula Weight 296.0007 g/mol