Learn More
(+)-Fenchone 100 μg/mL in Methanol, Dr. Ehrenstorfer
Product Code. 30317747
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30317747 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30317747 Supplier Dr. Ehrenstorfer Supplier No. DREA13460800ME100
Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material (+)-Fenchone
CAS 4695-62-9
Analyte or Component Names (+)-Fenchone
Shipping Condition +20°C
Molecular Formula C10 H16 O
Concentration 100 μg/mL
Certifications/Compliance ISO 17034
Synonym Bicyclo[2.2.1]heptan-2-one, 1,3,3-trimethyl-, (1S,4R)-, 2-Norbornanone, 1,3,3-trimethyl-, (1S,4R)-(+)- (8CI), Bicyclo[2.2.1]heptan-2-one, 1,3,3-trimethyl-, (1S)-, (+)-Fenchone, (1S)-Fenchone, (1S,4R)-1,3,3-Trimethylbicyclo[2.2.1]heptan-2-one, (S)-(+)-Fenchone, (S)-Fenchone, D(+)-Fenchone, D-Fenchone
InChI Formula InChI=1S/C10H16O/c1-9(2)7-4-5-10(3,6-7)8(9)11/h7H,4-6H2,1-3H3/t7-,10+/m1/s1
SMILES C[C@]12CC[C@H](C1)C(C)(C)C2=O
IUPAC Name (1R,4S)-2,2,4-trimethylbicyclo[2.2.1]heptan-3-one
Solvent or Matrix Methanol
Molecular Weight (g/mol) 152.23
Quantity 1 mL
Formula Weight 152.1201 g/mol